missing translation for 'onlineSavingsMsg'
Learn More

Trametinib Impurity-I, TRC

Product Code. 30425844
Change view
Click to view available options
Quantity:
2.5 mg
10 mg
Unit Size:
10mg
2.5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30425844 2.5 mg 2.5mg
30409059 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30425844 Supplier Toronto Research Chemicals Supplier No. TRCT1383402.5MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Trametinib Impurity-I
CAS 1383430-03-2
Molecular Formula C26 H23 F I N5 O5
Quantity 2.5 mg
Synonym Trametinib Impurity-I, Acetamide','N-[3-[3-cyclopropyl-5-[(2-fluoro-4-iodophenyl)imino]-3,4,5,6,7,8-hexahydro-8-hydroxy-6,8-dimethyl-2,4,7-trioxopyrido[4,3-d]pyrimidin-1(2H)-yl]phenyl]-, N-[3-[3-Cyclopropyl-5-[(2-fluoro-4-iodophenyl)imino]-3,4,5,6,7,8-hexahydro-8-hydroxy-6,8-di
InChI Formula InChI=1S/C26H23FIN5O5/c1-13(34)29-15-5-4-6-17(12-15)32-21-20(23(35)33(25(32)37)16-8-9-16)22(31(3)24(36)26(21,2)38)30-19-10-7-14(28)11-18(19)27/h4-7,10-12,16,38H,8-9H2,1-3H3,(H,29,34)
SMILES CN1C(=O)C(C)(O)C2=C(C(=O)N(C3CC3)C(=O)N2c4cccc(NC(=O)C)c4)C1=Nc5ccc(I)cc5F
IUPAC Name N-[3-[3-cyclopropyl-5-(2-fluoro-4-iodophenyl)imino-8-hydroxy-6,8-dimethyl-2,4,7-trioxopyrido[4,3-d]pyrimidin-1-yl]phenyl]acetamide
Molecular Weight (g/mol) 631.394
Formula Weight 631.073
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.