missing translation for 'onlineSavingsMsg'
Learn More

Trametinib (DMSO Solvate), TRC

Product Code. 30449466
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30449466 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30449466 Supplier Toronto Research Chemicals Supplier No. TRCT3143101G
 Restricted Item
Estimated Shipment: 21-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Trametinib Dimethyl Sulfoxide
CAS 1187431-43-1
Molecular Formula C26 H23 F I N5 O4 . C2 H6 O S
Quantity 1 g
Synonym Acetamide','N-[3-[3-cyclopropyl-5-[(2-fluoro-4-iodophenyl)amino]-3,4,6,7-tetrahydro-6,8-dimethyl-2,4,7-trioxopyrido[4,3-d]pyrimidin-1(2H)-yl]phenyl]-','compd. with 1,1'-sulfinylbis[methane] (1:1), GSK 1120212B, Trametinib dimethyl sulfoxide, GSK 1120212B
InChI Formula InChI=1S/C26H23FIN5O4.C2H6OS/c1-13-22-21(23(31(3)24(13)35)30-20-10-7-15(28)11-19(20)27)25(36)33(17-8-9-17)26(37)32(22)18-6-4-5-16(12-18)29-14(2)34;1-4(2)3/h4-7,10-12,17,30H,8-9H2,1-3H3,(H,29,34);1-2H3
SMILES CN1C(=O)C(=C2N(C(=O)N(C3CC3)C(=O)C2=C1Nc4ccc(I)cc4F)c5cccc(NC(=O)C)c5)C.CS(=O)C
IUPAC Name N-[3-[3-cyclopropyl-5-(2-fluoro-4-iodoanilino)-6,8-dimethyl-2,4,7-trioxopyrido[4,3-d]pyrimidin-1-yl]phenyl]acetamide;methylsulfinylmethane
Molecular Weight (g/mol) 693.53
Formula Weight 693.0918
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.