missing translation for 'onlineSavingsMsg'
Learn More

Trametinib (DMSO solvate), MedChemExpress

Product Code. 16404348
Encompass_Preferred
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
This item is not returnable. View return policy

Product Code. 16404348

Brand: Medchem Express HY10999A

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Trametinib (DMSO solvate) (GSK-1120212 (DMSO solvate);JTP-74057 (DMSO solvate)) is an orally active MEK inhibitor that inhibits MEK1 and MEK2 with IC50s of about 2 nM. Trametinib (DMSO solvate) activates autophagy and induces apoptosis.

Trametinib (DMSO solvate) (GSK-1120212 (DMSO solvate)

Specifications

Chemical Name or Material Trametinib (DMSO solvate)
CAS 1187431-43-1
Color Off-White
Purity Grade Notes Research
Molecular Formula C28H29FIN5O5S
Quantity 10 mg
Synonym GSK-1120212 (DMSO solvate) JTP-74057 (DMSO solvate)
Solubility Information DMSO : 69 mg/mL (99.49 mM; Need ultrasonic)
SMILES CN1C(C(C)=C(N(C(N(C2=O)C3CC3)=O)C4=CC=CC(NC(C)=O)=C4)C2=C1NC5=CC=C(C=C5F)I)=O.CS(C)=O
Molecular Weight (g/mol) 693.53
Formula Weight 693.53
Percent Purity 98.51%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.