missing translation for 'onlineSavingsMsg'
Learn More

Trametinib (DMSO solvate), MedChemExpress

Product Code. 16460667
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16460667 50 mg 50mg
16404348 10 mg 10mg
16475256 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16460667 Supplier Medchem Express Supplier No. HY10999A
 Restricted Item
Estimated Shipment: 18-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress Trametinib (DMSO solvate) (GSK-1120212 (DMSO solvate);JTP-74057 (DMSO solvate)) is an orally active MEK inhibitor that inhibits MEK1 and MEK2 with IC50s of about 2 nM. Trametinib (DMSO solvate) activates autophagy and induces apoptosis.

Trametinib (DMSO solvate) (GSK-1120212 (DMSO solvate)

Specifications

Chemical Name or Material Trametinib (DMSO solvate)
CAS 1187431-43-1
Color Off-White
Purity Grade Notes Research
Molecular Formula C28H29FIN5O5S
Quantity 50 mg
Synonym GSK-1120212 (DMSO solvate) JTP-74057 (DMSO solvate)
Solubility Information DMSO : 69 mg/mL (99.49 mM; Need ultrasonic)
SMILES CN1C(C(C)=C(N(C(N(C2=O)C3CC3)=O)C4=CC=CC(NC(C)=O)=C4)C2=C1NC5=CC=C(C=C5F)I)=O.CS(C)=O
Molecular Weight (g/mol) 693.53
Formula Weight 693.53
Percent Purity 98.51%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.