missing translation for 'onlineSavingsMsg'
Learn More

TRAM1 Antibody, Novus Biologicals™

Product Code. 18384179 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.05 mg
Unit Size:
0.05mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18384179 0.05 mg 0.05mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18384179 Supplier Novus Biologicals Supplier No. H00023471B01P

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Polyclonal Antibody

TRAM1 Polyclonal antibody specifically detects TRAM1 in Human samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen TRAM1
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1:500
Formulation PBS (pH 7.4)
Gene Accession No. AAH37738
Gene Alias PNAS8, PRO1292, TRAMP, TRAMtranslocating chain-associated membrane protein 1, translocating chain-associating membrane protein, translocation associated membrane protein 1, translocation-associating membrane protein 1
Host Species Mouse
Immunogen TRAM1 (AAH37738, 1 a.a. - 374 a.a.) full-length human protein. MAIRKKSTKSPPVLSHEFVLQNHADIVSCVAMVFLLGLMFEITAKASIIFVTLQYNVTLPATEEQATESVSLYYYGIKDLATVFFYMLVAIIIHAVIQEYMLDKINRRMHFSKTKHSKFNESGQLSAFYLFACVWGTFILISENYISDPTILWRAYPHNLMTFQMKFFYISQLAYWLHAFPELYFQKTKKEDIPRQLVYIGLYLFHIAGAYLLNLNHLGLVLLVLHYFVEFLFHISRLFYFSNEKYQKGFSLWAVLFVLGRLLTLILSVLTVGFGLARAENQKLDFSTGNFNVLAVRIAVLASICVTQAFMMWKFINFQLRRWREHSAFQAPAVKKKPTVTKGRSSKKGTENGVNGTLTSNVADSPRNKKEKSS
Purification Method Protein A purified
Quantity 0.05 mg
Regulatory Status RUO
Research Discipline Immunology, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 23471
Target Species Human
Content And Storage Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Mostrar más Mostrar menos
Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.