missing translation for 'onlineSavingsMsg'
Learn More

TRAM-34, MedChemExpress

Product Code. 16415728
Encompass_Preferred
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
This item is not returnable. View return policy

Product Code. 16415728

Brand: Medchem Express HY13519

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress TRAM-34 is a highly selective blocker of intermediate-conductance calcium-activated K+ channel (IKCa1) (Kd=20 nM).

TRAM-34 is a highly selective blocker of intermediate-conductance calcium-activated K+ channel (IKCa1) (Kd=20 nM).

Specifications

Chemical Name or Material TRAM-34
Physical Form Powder
CAS 289905-88-0
Color White
Quantity 10 mg
Purity Grade Notes Research
Molecular Formula C22H17ClN2
For Use With (Application) Neuroscience-Neuromodulation
Solubility Information DMSO : 25 mg/mL (72.50 mM; Need ultrasonic)
SMILES ClC1=CC=CC=C1C(N2N=CC=C2)(C3=CC=CC=C3)C4=CC=CC=C4
Molecular Weight (g/mol) 344.84
Formula Weight 344.84
Percent Purity 99.32%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.