missing translation for 'onlineSavingsMsg'
Learn More

Tralomethrin-d5 (>90%), TRC

Product Code. 30457217
Change view
Click to view available options
Quantity:
1 mg
10 mg
Unit Size:
10mg
1mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30457217 10 mg 10mg
30455102 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30457217 Supplier Toronto Research Chemicals Supplier No. TRCT71240210MG
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Tralomethrin-d5
Molecular Formula C22H14D5Br4NO3
Quantity 10 mg
Synonym 2,2-Dimethyl-3-(1,2,2,2-tetrabromoethyl)cyclopropanecarboxylic Acid Cyano(3-phenoxyphenyl)methyl Ester-d5, Bengal Fire Ant Killer-d5, HAG 107-d5, RU 25472-d5, SAGA-d5, Scout-d5, Scout (insecticide)-d5, Scout X-tra-d5, Tracker-d5
SMILES O=C(C1C(C)(C)C1C(C(Br)(Br)Br)Br)OC(C#N)C2=CC(OC3=C([2H])C([2H])=C([2H])C([2H])=C3[2H])=CC=C2
Molecular Weight (g/mol) 670.04
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.