Learn More
TRAK2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 13770-1-AP-20UL
 View more versions of this product
Product Code. 16852154
  • £135.00 / 20µL
Description
May regulate endosome-to-lysosome trafficking of membrane cargo, including EGFR.
Specifications
Specifications
TRAK2
Polyclonal
Unconjugated
TRAK2
ALS2CR3, CALS C, GRIF 1, GRIF1, KIAA0549, OIP98, TRAK2
Rabbit
Antigen Affinity Chromatography
RUO
171086, 66008, 70827
-20°C
Liquid
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
0.28 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
O60296, Q8R2H7
Trak2
TRAK2 Fusion Protein Ag4753
20 μL
Primary
Human, Rat, Mouse