missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ TRAK2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA534889
This item is not returnable.
View return policy
Description
Recommended positive controls: mouse brain. Predicted reactivity: Human (99%), Mouse (90%), Rat (88%), Rhesus Monkey (99%), Bovine (91%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
May regulate endosome-to-lysosome trafficking of membrane cargo, including EGFR.
Specifications
| TRAK2 | |
| Polyclonal | |
| Unconjugated | |
| TRAK2 | |
| 2900022D04Rik; 4733401O11Rik; AI480836; Als2cr3; amyotrophic lateral sclerosis 2 (juvenile) chromosome region candidate 3; amyotrophic lateral sclerosis 2 (juvenile) chromosome region, candidate 3; amyotrophic lateral sclerosis 2 (juvenile) chromosome region, candidate 3 homolog; amyotrophic lateral sclerosis 2 chromosomal region candidate gene 3 protein; Amyotrophic lateral sclerosis 2 chromosomal region candidate gene 3 protein homolog; AW046271; CALS-C; EMBL AAH60681.1; GABA-A receptor interacting factor-1; GABA-A receptor-interacting factor 1; gamma-aminobutyric acid(A) receptor-interacting factor; Grif1; GRIF-1; KIAA0549; MILT2; milton homolog 2; mKIAA0549; O-GlcNAc transferase-interacting protein of 98 kDa; OIP98; O-linked N-acetylglucosamine transferase interacting protein 98; trafficking kinesin protein 2; trafficking kinesin-binding protein 2; trafficking protein, kinesin binding 2; Trak2 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 66008, 70827 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 1% BSA, 20% glycerol and 0.01% thimerosal; pH 7 | |
| O60296 | |
| TRAK2 | |
| Recombinant protein encompassing a sequence within the C-terminus region of human TRAK2. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction