missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ TRAK2 Polyclonal Antibody

Goat Polyclonal Antibody
Brand: Invitrogen™ PA5142039
This item is not returnable.
View return policy
Description
This antibody is tested in Peptide ELISA: antibody detection limit dilution 1:32,000.
May regulate endosome-to-lysosome trafficking of membrane cargo, including EGFR.
Specifications
| TRAK2 | |
| Polyclonal | |
| Unconjugated | |
| TRAK2 | |
| 2900022D04Rik; 4733401O11Rik; AI480836; Als2cr3; amyotrophic lateral sclerosis 2 (juvenile) chromosome region candidate 3; amyotrophic lateral sclerosis 2 (juvenile) chromosome region, candidate 3; amyotrophic lateral sclerosis 2 (juvenile) chromosome region, candidate 3 homolog; amyotrophic lateral sclerosis 2 chromosomal region candidate gene 3 protein; Amyotrophic lateral sclerosis 2 chromosomal region candidate gene 3 protein homolog; AW046271; CALS-C; EMBL AAH60681.1; GABA-A receptor interacting factor-1; GABA-A receptor-interacting factor 1; gamma-aminobutyric acid(A) receptor-interacting factor; Grif1; GRIF-1; KIAA0549; MILT2; milton homolog 2; mKIAA0549; O-GlcNAc transferase-interacting protein of 98 kDa; OIP98; O-linked N-acetylglucosamine transferase interacting protein 98; trafficking kinesin protein 2; trafficking kinesin-binding protein 2; trafficking protein, kinesin binding 2; Trak2 | |
| Goat | |
| Ammonium sulfate precipitation | |
| RUO | |
| 66008 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Flow Cytometry, Immunocytochemistry | |
| 0.5 mg/mL | |
| TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
| O60296 | |
| TRAK2 | |
| Peptide with sequence CTSSPKMGVLKED (aa 902-914). | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction