missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TRAILR4/TNFRSF10D/DcR2 Rabbit anti-Human, Clone: 9, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP2-89478-50UL
This item is not returnable.
View return policy
Description
TRAILR4/TNFRSF10D/DcR2 Monoclonal specifically detects TRAILR4/TNFRSF10D/DcR2 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| TRAILR4/TNFRSF10D/DcR2 | |
| Monoclonal | |
| Unconjugated | |
| 0.2 um filtered solution in PBS | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| 9 | |
| Immunohistochemistry, Immunohistochemistry-Paraffin 1:100-1:500 | |
| CD264, CD264 antigen, DcR2, DCR2decoy with truncated death domain, Decoy receptor 2, TNF receptor-related receptor for TRAIL, TNF-related apoptosis-inducing ligand receptor 4, TRAIL-R4, TRAILR4TRAIL receptor 4, TRUNDDTRAIL receptor with a truncated death domain, tumor necrosis factor receptor superfamily member 10D, tumor necrosis factor receptor superfamily, member 10d, decoy with truncateddeath domain | |
| Recombinant Human TRAILR4/TNFRSF10D/DcR2 protein | |
| 50 μg | |
| Apoptosis, Cancer, Cell Biology, Cytokine Research, Immunology, Innate Immunity, Phospho Specific, Signal Transduction | |
| 8793 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction