missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TRAILR4/TNFRSF10D/DcR2 Antibody (102926) - Azide and BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
£167.00 - £396.00
Specifications
| Antigen | TRAILR4/TNFRSF10D/DcR2 |
|---|---|
| Clone | 102926 |
| Concentration | 1.0 mg/mL |
| Dilution | Blocking/Neutralizing |
| Applications | Blocking Assay |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18042444
|
Novus Biologicals
MAB6331-SP |
25 μg |
£167.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18083606
|
Novus Biologicals
MAB6331 |
100 μg |
£396.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
TRAILR4/TNFRSF10D/DcR2 Monoclonal specifically detects TRAILR4/TNFRSF10D/DcR2 in Human samples. It is validated for Block/Neutralize.Specifications
| TRAILR4/TNFRSF10D/DcR2 | |
| 1.0 mg/mL | |
| Blocking Assay | |
| Unconjugated | |
| Mouse | |
| Apoptosis, Cancer, Cell Biology, Cytokine Research, Immunology, Innate Immunity | |
| PBS with No Preservative | |
| 8793 | |
| Partial recombinant human TRAIL R4 protein (amino acids 56-211) [UniProt Q9UBN6] | |
| Primary | |
| Store at -20C. Avoid freeze-thaw cycles. |
| 102926 | |
| Blocking/Neutralizing | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| CD264, CD264 antigen, DcR2, DCR2decoy with truncated death domain, Decoy receptor 2, TNF receptor-related receptor for TRAIL, TNF-related apoptosis-inducing ligand receptor 4, TRAIL-R4, TRAILR4TRAIL receptor 4, TRUNDDTRAIL receptor with a truncated death domain, tumor necrosis factor receptor superfamily member 10D, tumor necrosis factor receptor superfamily, member 10d, decoy with truncateddeath domain | |
| TNFRSF10D | |
| IgG1 | |
| Protein A or G purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title