missing translation for 'onlineSavingsMsg'
Learn More

TRAILR1/TNFRSF10A Rabbit anti-Human, Clone: 101, Novus Biologicals™

Rabbit Monoclonal Antibody

£296.00 - £429.00

Specifications

Antigen TRAILR1/TNFRSF10A
Clone 101
Dilution Flow Cytometry 1:25-1:100
Applications Flow Cytometry
Classification Monoclonal
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18224107
View Documents
Novus Biologicals
NBP2-89476-50UL
50 μg
£296.00
50µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18244847
View Documents
Novus Biologicals
NBP2-89476-100UL
100 μg
£429.00
100µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

TRAILR1/TNFRSF10A Monoclonal specifically detects TRAILR1/TNFRSF10A in Human samples. It is validated for Flow Cytometry.
Specifications

Specifications

TRAILR1/TNFRSF10A
Flow Cytometry 1:25-1:100
Monoclonal
Purified
RUO
Human
APO2, CD261, CD261 antigen, cytotoxic TRAIL receptor, Death receptor 4, DR4, DR4 TRAIL receptor 1, TNF-related apoptosis inducing ligand receptor 1, TNF-related apoptosis-inducing ligand receptor 1, TRAIL-R1, TRAILR-1, TRAILR1MGC9365, tumor necrosis factor receptor superfamily member 10A, tumor necrosis factor receptor superfamily, member 10a
Recombinant Human TRAILR1/TNFRSF10A Protein
Primary
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
101
Flow Cytometry
Unconjugated
Rabbit
Apoptosis, Cancer
0.2 um filtered solution in PBS
8797
IgG
Protein A purified
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.