missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ TRAILR1/TNFRSF10A Antibody Pair [HRP]

Product Code. 18240047 Shop All Bio Techne Products
Click to view available options
Quantity:
15 Plates
5 Plates
Unit Size:
Pack of 15
Pack of 5
This item is not returnable. View return policy

Product Code. 18240047

Brand: Novus Biologicals™ NBP27959915PLATES

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Monoclonal Antibody Pair

This solid phase sandwich ELISA utilizes a monoclonal antibody specific for Human TRAILR1/TNFRSF10A.
TRUSTED_SUSTAINABILITY

Specifications

Assay Range 2.34 pg/mL
Assay Sensitivity 2.34-150 pg/mL
Assay Sandwich ELISA
Conjugate HRP
Immunoassay Kit Format Sandwich ELISA Kit
Product Type Antibody Pairs
Specificity This solid phase sandwich ELISA utilizes a monoclonal antibody specific for Human TRAILR1/TNFRSF10A.
Target Species Human
For Use With (Application) Sandwich ELISA
Gene Alias APO2, CD261, CD261 antigen, cytotoxic TRAIL receptor, Death receptor 4, DR4, DR4 TRAIL receptor 1, TNF-related apoptosis inducing ligand receptor 1, TNF-related apoptosis-inducing ligand receptor 1, TRAIL-R1, TRAILR-1, TRAILR1MGC9365, tumor necrosis factor receptor superfamily member 10A, tumor necrosis factor receptor superfamily, member 10a
Gene ID (Entrez) 8797
Gene Symbol TNFRSF10A
Host Species Rabbit/Mouse
Kit Contents
  • Detection Antibody: 0.2 mg/mL of rabbit anti-Human TRAILR1/TNFRSF10A monoclonal antibody conjugated to horseradish-peroxidase (HRP) (in PBS, 50 % HRP-Protector, pH 7.4, store at 4C).
  • Capture Antibody: 0.5 mg/mL of mouse anti-Human TRAILR1/TNFRSF10A monoclonal antibody (in PBS, pH 7.4).
  • Standard: Each vial contains 30 ng of recombinant Human TRAILR1/TNFRSF10A.
  • Quantity 15 Plates
    Regulatory Status RUO
    Research Discipline Apoptosis, Cancer
    Storage Requirements Storage is content dependent.
    Target TRAILR1/TNFRSF10A
    Technique 1. Capture Antibody: Dilute to a working concentration of 2 μg/mL in PBS before coating. 2. Detection Antibody: Dilute to working concentration of 0.25 μg/mL in detection antibody dilution buffer before use. 3. Standard: Reconstitute with 1 ml detection antibody dilution buffer. After reconstitution, store at -20 to -80°C in a manual defrost freezer. A seven-point standard curve using 2-fold serial dilutions in sample dilution buffer, and a high standard of 150 pg/mL is recommended.
    Show More Show Less
    Product Content Correction

    Your input is important to us. Please complete this form to provide feedback related to the content on this product.

    Product Title

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

    Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.