missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TRAILR1/TNFRSF10A Antibody (DR-4-02), FITC, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-31343F
This item is not returnable.
View return policy
Description
TRAILR1/TNFRSF10A Monoclonal specifically detects TRAILR1/TNFRSF10A in Human samples. It is validated for Flow Cytometry, Immunocytochemistry/Immunofluorescence.
Specifications
| TRAILR1/TNFRSF10A | |
| Monoclonal | |
| FITC | |
| PBS with 0.05% Sodium Azide | |
| TNFRSF10A | |
| Fusion protein (Human). Fusion protein containing the extracellular part of DR4 and the constant part of the heavy chain of the human IgG1 (NM_003844.3). | |
| 0.1 mL | |
| Apoptosis, Cancer | |
| 8797 | |
| Human | |
| Purified |
| Flow Cytometry, Immunocytochemistry, Immunofluorescence | |
| DR-4-02 | |
| Flow Cytometry, Immunocytochemistry/Immunofluorescence | |
| APO2, CD261, CD261 antigen, cytotoxic TRAIL receptor, Death receptor 4, DR4, DR4 TRAIL receptor 1, TNF-related apoptosis inducing ligand receptor 1, TNF-related apoptosis-inducing ligand receptor 1, TRAIL-R1, TRAILR-1, TRAILR1MGC9365, tumor necrosis factor receptor superfamily member 10A, tumor necrosis factor receptor superfamily, member 10a | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| The mouse monoclonal antibody DR-4-02 recognizes an extracellular epitope of TRAIL-R1 (DR4), a human death receptor 4 expressed in most human tissues (spleen, peripheral blood leucocytes, thymus) and in a variety of tumour-derived cell lines. | |
| Store at 4C in the dark. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction