missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TRAILR1/TNFRSF10A Antibody (B-N36), DyLight 755, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-14599IR
This item is not returnable.
View return policy
Description
TRAILR1/TNFRSF10A Monoclonal antibody specifically detects TRAILR1/TNFRSF10A in Human samples. It is validated for Flow Cytometry, ELISA
Specifications
| TRAILR1/TNFRSF10A | |
| Monoclonal | |
| DyLight 755 | |
| APO2, CD261, CD261 antigen, cytotoxic TRAIL receptor, Death receptor 4, DR4, DR4 TRAIL receptor 1, TNF-related apoptosis inducing ligand receptor 1, TNF-related apoptosis-inducing ligand receptor 1, TRAIL-R1, TRAILR-1, TRAILR1MGC9365, tumor necrosis factor receptor superfamily member 10A, tumor necrosis factor receptor superfamily, member 10a | |
| Recombinant human TRAILR1/TNFRSF10A | |
| 0.1 mL | |
| Apoptosis, Cancer | |
| 8797 | |
| Store at 4°C in the dark. | |
| IgG1 κ |
| Flow Cytometry, ELISA | |
| B-N36 | |
| 50mM Sodium Borate | |
| Mouse | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction