missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TRAIL/TNFSF10 Mouse anti-Human, Clone: B-T24, Novus Biologicals™
Mouse Monoclonal Antibody
£319.00 - £603.00
Specifications
| Antigen | TRAIL/TNFSF10 |
|---|---|
| Clone | B-T24 |
| Concentration | 1 mg/mL |
| Dilution | Flow Cytometry, ELISA, Functional |
| Applications | Flow Cytometry, ELISA, Functional Assay |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18344647
|
Novus Biologicals
NBP3-14605-200UL |
200 μg |
£319.00
200µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18336536
|
Novus Biologicals
NBP3-14605-500UL |
500 μg |
£603.00
500µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
TRAIL/TNFSF10 Monoclonal antibody specifically detects TRAIL/TNFSF10 in Human samples. It is validated for Flow Cytometry, ELISA, Functional AssaySpecifications
| TRAIL/TNFSF10 | |
| 1 mg/mL | |
| Flow Cytometry, ELISA, Functional Assay | |
| Unconjugated | |
| Mouse | |
| Apoptosis, Cancer, Cell Biology, Death Receptor Signaling Pathway, Signal Transduction, Tumor Suppressors | |
| PBS. Sterile-filtered through 0.22 um. | |
| 8743 | |
| IgG1 κ | |
| Affinity purified | |
| Antibody |
| B-T24 | |
| Flow Cytometry, ELISA, Functional | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| Apo-2 ligand, APO2L, Apo-2Ltumor necrosis factor (ligand) family, member 10, APO2Ltumor necrosis factor apoptosis-inducing ligand splice variant delta, CD253, CD253 antigen, Protein TRAIL, TL2, TNF-related apoptosis-inducing ligand, TRAILTNF-related apoptosis inducing ligand TRAIL, tumor necrosis factor (ligand) superfamily, member 10, tumor necrosis factor ligand superfamily member 10 | |
| Recombinant human TRAIL/TNFSF10 | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Titolo del prodotto