missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TRAIL/TNFSF10 Antibody (55B709.3) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody has been used in 8 publications
£189.00 - £417.00
Specifications
| Antigen | TRAIL/TNFSF10 |
|---|---|
| Clone | 55B709.3 |
| Concentration | 1.0 mg/mL |
| Dilution | Western Blot 1-2 ug/ml, Immunohistochemistry-Paraffin 5 ug/ml |
| Applications | Western Blot, Immunohistochemistry (Paraffin) |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18054885
|
Novus Biologicals
NB100-56518SS |
0.025 mg |
£189.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18727703
|
Novus Biologicals
NB100-56518 |
0.1 mg |
£417.00
0.10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
TRAIL/TNFSF10 Monoclonal specifically detects TRAIL/TNFSF10 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
| TRAIL/TNFSF10 | |
| 1.0 mg/mL | |
| Western Blot, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Mouse | |
| Apoptosis, Cancer, Cell Biology, Death Receptor Signaling Pathway, Signal Transduction, Tumor Suppressors | |
| NP_003801.1, P50592 | |
| 8743 | |
| This monoclonal antibody was raised against a peptide corresponding to amino acids 17-35 of human TRAIL. This peptide sequence has been shown to be involved in TRAIL and DR5 (TRAIL-R2) interaction. | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| 55B709.3 | |
| Western Blot 1-2 ug/ml, Immunohistochemistry-Paraffin 5 ug/ml | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| Apo-2 ligand, APO2L, Apo-2Ltumor necrosis factor (ligand) family, member 10, APO2Ltumor necrosis factor apoptosis-inducing ligand splice variant delta, CD253, CD253 antigen, Protein TRAIL, TL2, TNF-related apoptosis-inducing ligand, TRAILTNF-related apoptosis inducing ligand TRAIL, tumor necrosis factor (ligand) superfamily, member 10, tumor necrosis factor ligand superfamily member 10 | |
| TNFSF10 | |
| IgG1 κ | |
| Protein G purified | |
| In Jurkat, a 32 kDa band, which represents a membrane bound form of TRAIL, is observed. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title