missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ TRAIL-R2 (DR5) Chimeric Recombinant Rabbit Monoclonal Antibody (304)

Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA551601
This item is not returnable.
View return policy
Description
The antibody is specific to human DR5.
DR5 (Apo2, TRAIL-R2, TRICK2, KILLER) is a recently identified death domain containing receptor for TRAIL, which mediates TRAIL induced apoptosis. DR5 is a member of the TNF-receptor superfamily, and contains an intracellular death domain. DR5 can be activated by tumor necrosis factor-related apoptosis inducing ligand (TNFSF10/TRAIL/APO-2L), and transduces an apoptosis signal. Studies with FADD-deficient mice suggested that FADD, a death domain containing adaptor protein, is required for the apoptosis mediated by DR5. Two transcript variants encoding different isoforms of DR5 and one non-coding transcript have been found. Overexpression of DR5 induces apoptosis and activates NF-kappa-B. DR5 is expressed in a number of cell types, and to particularly high levels in lymphocytes and spleen. Diseases associated with DR5 dysfunction include squamous cell carcinoma, and diffuse infiltrative lymphocytosis syndrome.
Specifications
| TRAIL-R2 (DR5) Chimeric | |
| Recombinant Monoclonal | |
| 1 mg/mL | |
| PBS with 0.02% ProClin 300 | |
| O14763 | |
| TNFRSF10B | |
| The antibody was raised in mice homozygous for both disrupted endogenous Ig heavy chain and k light chain, harboring a chromosome 14 fragment (SC20) containing a human Ig heavy chain locus and a human Igk chain transgene (KCo5). The mice were immunised against TRAIL-R1 and TRAIL-R2. | |
| 200 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG κ |
| ELISA, Flow Cytometry, Functional Assay, Radioimmune Assays (RIA), Western Blot, Immunocytochemistry | |
| 304 | |
| Unconjugated | |
| TNFRSF10B | |
| apoptosis inducing protein TRICK2A/2B; apoptosis inducing receptor TRAIL-R2; CD262; cytotoxic TRAIL receptor-2; death domain containing receptor for TRAIL/Apo-2L; death receptor 5; Dr5; dr-5; Fas-like protein; KILLER; KILLER/DR5; KILLER/DR5 TRAIL death-inducing receptor; Ly98; MK; p53-regulated DNA damage-inducible cell death receptor(killer); sCD262; soluble CD262; TNF receptor superfamily member 10b; TNF-related apoptosis-inducing ligand receptor 2; TNFRSF10B; TRAIL receptor 2; TRAILR2; TRAIL-R2; TRICK2; TRICK2A; TRICK2B; TRICKB; tumor necrosis factor receptor superfamily member 10B; tumor necrosis factor receptor superfamily, member 10b; tumor necrosis factor receptor-like protein ZTNFR9; UNQ160/PRO186; ZTNFR9 | |
| Rabbit | |
| Protein A | |
| RUO | |
| 8795 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction