Learn More
TRAFD1 Rabbit anti-Human, Unconjugated, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 27741-1-AP-150UL
This item is not returnable.
View return policy
Description
The TRAFD1 antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human TRAFD1. This antibody recognizes human antigen. The TRAFD1 antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
| TRAFD1 | |
| Polyclonal | |
| Unconjugated | |
| 150 μL | |
| 10906 | |
| IgG |
| Western Blot, ELISA | |
| 200 μg/mL | |
| Rabbit | |
| Primary | |
| Human |