missing translation for 'onlineSavingsMsg'
Learn More

TRAFD1 Antibody (OTI3E8) [Alexa Fluor™ 750], Novus Biologicals™

Product Code. 30687949 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30687949 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30687949 Supplier Novus Biologicals Supplier No. NBP344943AF750

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

TRAFD1 Monoclonal antibody specifically detects TRAFD1 in Human samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen TRAFD1
Applications Western Blot
Classification Monoclonal
Clone OTI3E8
Conjugate Alexa Fluor 750
Formulation 50mM Sodium Borate
Gene Alias FLN29FLN29 gene product, Protein FLN29, TRAF-type zinc finger domain containing 1, TRAF-type zinc finger domain-containing protein 1
Host Species Mouse
Immunogen Human recombinant protein fragment corresponding to amino acids 308-582 of human TRAFD1(NP_006691) produced in E.coli.
Purification Method >95%
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Immunology, Innate Immunity
Primary or Secondary Primary
Gene ID (Entrez) 10906
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.