missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TRAFD1 Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-94278-0.1ml
This item is not returnable.
View return policy
Description
TRAFD1 Polyclonal antibody specifically detects TRAFD1 in Human samples. It is validated for Western Blot
Specifications
| TRAFD1 | |
| Polyclonal | |
| Western Blot 1:500-1:1000 | |
| FLN29FLN29 gene product, Protein FLN29, TRAF-type zinc finger domain containing 1, TRAF-type zinc finger domain-containing protein 1 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-260 of human TRAFD1 (NP_006691.1). MAEFLDDQETRLCDNCKKEIPVFNFTIHEIHCQRNIGMCPTCKEPFPKSDMETHMAAEHCQVTCKCNKKLEKRLLKKHEETECPLRLAVCQHCDLELSILKLKEHEDYCGARTELCGNCGRNVLVKDLKTHPEVCGREGEEKRNEVAIPPNAYDESWGQDGIWIASQLLRQIEALDPPMRLPRRPLRAFESDVFHNRTTNQRNITAQVSIQNNLFEEQERQERNRGQQPPKEGGEESANLDFMLALSLQNEGQASSVAEQ | |
| 0.1 mL | |
| Immunology, Innate Immunity | |
| 10906 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction