missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TRAF7 Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
£149.00 - £366.00
Specifications
| Antigen | TRAF7 |
|---|---|
| Dilution | Western Blot 1:500-1:2000 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18696002
|
Novus Biologicals
NBP2-94409-0.02ml |
0.02 mL |
£149.00
0.02mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18610222
|
Novus Biologicals
NBP2-94409-0.1ml |
0.1 mL |
£366.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
TRAF7 Polyclonal antibody specifically detects TRAF7 in Rat samples. It is validated for Western BlotSpecifications
| TRAF7 | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Lipid and Metabolism, Zinc Finger | |
| PBS (pH 7.3), 50% glycerol | |
| 84231 | |
| IgG | |
| Affinity purified |
| Western Blot 1:500-1:2000 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Rat | |
| DKFZp586I021, E3 ubiquitin-protein ligase TRAF7, EC 6.3.2.-, MGC7807, RFWD1RING finger protein 119, RNF119RING finger and WD repeat-containing protein 1, TNF receptor-associated factor 7ring finger and WD repeat domain 1 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 260-400 of human TRAF7 (NP_115647.2). PHSKYGCTFIGNQDTYETHLETCRFEGLKEFLQQTDDRFHEMHVALAQKDQEIAFLRSMLGKLSEKIDQLEKSLELKFDVLDENQSKLSEDLMEFRRDASMLNDELSHINARLNMGILGSYDPQQIFKCKGTFVGHQGPVW | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title