missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ TRAF5 Monoclonal Antibody (OTI7A3)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA527289
This item is not returnable.
View return policy
Description
TRAF5 Monoclonal Antibody for Western Blot
Tumor necrosis factor (TNF) induced signaling is mediated through association of TNF receptor (TNFR) with adaptor proteins, such as TNF receptor associated proteins (TRAFs). TRAFs form a family of cytoplasmic adapter proteins that mediate signal transduction from many members of the TNF-receptor superfamily (e. g. RANK, CD30, CD40, etc. ) and the interleukin-1 receptor. The carboxy-terminal region of TRAFs is required for self-association and interaction with receptor cytoplasmic domains following ligand-induced oligomerization. Recent molecular cloning studies have lead to identification of six TRAFs (TRAF1-TRAF6) (1-4). TRAF5 is a 558-amino acid protein. TRAF5 is implicated in NF-kB and c-Jun NH(2)-terminal kinase/stress-activated protein kinase activation by members of the TNF receptor superfamily, including CD27, CD30, CD40, and lymphotoxin-beta receptor. Targeted disruption of TRAF5 gene causes defects in CD40-CD27 mediated lymphocyte activation (5).
Specifications
| TRAF5 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| O00463 | |
| TRAF5 | |
| Human recombinant protein fragment corresponding to amino acids 334-557 of TRAF5 produced in E.coli. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Western Blot | |
| OTI7A3 | |
| Unconjugated | |
| TRAF5 | |
| 2700049M22Rik; AI875481; AU022477; BH3 interacting domain death agonist; BH3-interacting domain death agonist; BH3-interacting domain death agonist p11; BH3-interacting domain death agonist p13; BH3-interacting domain death agonist p15; BID; MGC:39780; p11 BID; p13 BID; p15 BID; p22 BID; RING finger protein 84; RNF84; TNF receptor associated factor 5; TNF receptor-associated factor 5; Traf5 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 7188 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction