missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TRAF3IP3 Rabbit Polyclonal Antibody, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 18110-1-AP-20UL
This item is not returnable.
View return policy
Description
The TRAF3IP3 antibody from Proteintech is a rabbit polyclonal antibody generated with fusion protein of human TRAF3IP3. This antibody recognizes human, mouse antigen. The TRAF3IP3 antibody has been validated for the following applications: WB, IHC, ELISA analysis.Specifications
| TRAF3IP3 | |
| Polyclonal | |
| Unconjugated | |
| Rabbit | |
| Antigen affinity purification | |
| RUO | |
| 80342 | |
| Store at -20°C. Stable for one year after shipment. | |
| Polyclonal Antibody | |
| IgG |
| Western Blot, Immunohistochemistry, ELISA | |
| 1000 μg/mL | |
| DJ434O14.3; FLJ44151; MGC117354; MGC163289; T3JAM | |
| Fusion Protein | |
| 20 μL | |
| Primary | |
| Human, Mouse | |
| 64 kDa, 47 kDa, 41 kDa | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction