missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ TRAF3IP3 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA543460
This item is not returnable.
View return policy
Description
Peptide sequence: KQKMVILQDL LSTLIQASDS SWKGQLNEDK LKGKLRSLEN QLYTCTQKYS Sequence homology: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%.
The gene encodes a protein that mediates cell growth by modulating the c-Jun N-terminal kinase signal transduction pathway. The encoded protein may also interact with a large multi-protein assembly containing the phosphatase 2A catalytic subunit. Alternative splicing results in multiple transcript variants. [provided by RefSeq, Dec 2013].
Specifications
| TRAF3IP3 | |
| Polyclonal | |
| Unconjugated | |
| TRAF3IP3 | |
| 6030423D04Rik; AI115021; DJ434O14.3; FLJ44151; MGC117354; MGC163289; RGD1304848; T3JAM; TRAF3 interacting Jun N terminal kinase (JNK) activating modulator; TRAF3 interacting protein 3; TRAF3-interacting JNK-activating modulator; TRAF3-interacting JNK-activating modulator-like protein; TRAF3-interacting Jun N-terminal kinase (JNK)-activating modulator; TRAF3-interacting protein 3; Traf3ip3 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 80342 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q9Y228 | |
| TRAF3IP3 | |
| Synthetic peptide directed towards the middle region of human TRAF3IP3. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction