missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TRAF3IP2 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 26692-1-AP-20UL
This item is not returnable.
View return policy
Description
This gene encodes a protein involved in regulating responses to cytokines by members of the Rel/NF-kappaB transcription factor family. These factors play a central role in innate immunity in response to pathogens, inflammatory signals and stress. This gene product interacts with TRAF proteins (tumor necrosis factor receptor-associated factors) and either I-kappaB kinase or MAP kinase to activate either NF-kappaB or Jun kinase. Two alternative transcripts encoding different proteins have been identified. A third transcript, which does not encode a protein and is transcribed in the opposite orientation, has been identified. Overexpression of this transcript has been shown to reduce expression of at least one of the protein encoding transcripts, suggesting it has a regulatory role in the expression of this gene.Specifications
| TRAF3IP2 | |
| Polyclonal | |
| Unconjugated | |
| TRAF3IP2 | |
| ACT1, Adapter protein CIKS, C6orf2, C6orf4, C6orf5, C6orf6, CIKS, Connection to IKK and SAPK/JNK, TRAF3 interacting protein 2, TRAF3IP2 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 10758 | |
| -20°C | |
| Liquid |
| Western Blot, Immunocytochemistry, Immunofluorescence | |
| 0.47 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| O43734 | |
| TRAF3IP2 | |
| TRAF3IP2 Fusion Protein Ag24840 | |
| 20 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction