missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ TRAF3IP2 Polyclonal Antibody
GREENER_CHOICE

Product Code. 12838202
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
12838202 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 12838202 Supplier Invitrogen™ Supplier No. PA519988

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

In Western blot applications, this antibody detects a band at ~63kDa. A suggested positive control is human placenta lysate. PA5-19988 can be used with blocking peptide PEP-0112.

This gene encodes a protein involved in regulating responses to cytokines by members of the Rel/NF-kappaB transcription factor family. These factors play a central role in innate immunity in response to pathogens, inflammatory signals and stress. This gene product interacts with TRAF proteins (tumor necrosis factor receptor-associated factors) and either I-kappaB kinase or MAP kinase to activate either NF-kappaB or Jun kinase. Two alternative transcripts encoding different proteins have been identified. A third transcript, which does not encode a protein and is transcribed in the opposite orientation, has been identified. Overexpression of this transcript has been shown to reduce expression of at least one of the protein encoding transcripts, suggesting it has a regulatory role in the expression of this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TRAF3IP2
Applications Immunohistochemistry, Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.02% sodium azide
Gene TRAF3IP2
Gene Accession No. O43734
Gene Alias ACT1; adapter protein CIKS; AI429613; C6orf2; C6orf4; C6orf5; C6orf6; CANDF8; CIKS; connection to IKK and SAPK/JNK; NFkB-activating protein ACT1; nuclear factor NF-kappa-B activator 1; PSORS13; RGD1304768; RP3-487J7.3; TRAF3 interacting protein 2; TRAF3-interacting protein 2; TRAF3IP2
Gene Symbols TRAF3IP2
Host Species Rabbit
Immunogen A synthetic peptide corresponding to amino acids 554 to 568 of human CIKS.
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10758
Target Species Human
Content And Storage Maintain refrigerated at 2-8°C for up to 3 months. For long term storage store at -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.