missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TRAF3IP2 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NB100-93556
This item is not returnable.
View return policy
Description
TRAF3IP2 Polyclonal specifically detects TRAF3IP2 in Human, Mouse samples. It is validated for Western Blot.
Specifications
| TRAF3IP2 | |
| Polyclonal | |
| Unconjugated | |
| PBS with 0.02% Sodium Azide | |
| TRAF3IP2 | |
| Synthetic peptide corresponding to the N-terminus of human ACT. | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Reacts with the N terminus of ACT | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| Western Blot | |
| 1 mg/mL | |
| Western Blot 1:100-1:2000 | |
| ACT1DKFZP586G0522, adapter protein CIKS, C6orf2, C6orf4chromosome 6 open reading frame 5, C6orf5DKFZp586G0522, C6orf6MGC3581, CIKSchromosome 6 open reading frame 2, Connection to IKK and SAPK/JNK, NFkB-activating protein ACT1, Nuclear factor NF-kappa-B activator 1, TRAF3 interacting protein 2, TRAF3-interacting protein 2 | |
| Rabbit | |
| 55 kDa | |
| 0.1 mg | |
| Signal Transduction | |
| 10758 | |
| Human, Mouse | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction