missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TRAF3 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 18099-1-AP-20UL
This item is not returnable.
View return policy
Description
Tumor necrosis factor (TNF) induced signaling is mediated through association of TNF receptor (TNFR) with adaptor proteins, such as TNF receptor associated proteins (TRAFs). TRAFs form a family of cytoplasmic adapter proteins that mediate signal transduction from many members of the TNF-receptor superfamily (e.g. RANK, CD30, CD40, etc.) and the interleukin-1 receptor. The carboxy-terminal region of TRAFs is required for self-association and interaction with receptor cytoplasmic domains following ligand-induced oligomerization. Recent molecular cloning studies have lead to identification of six TRAFs (TRAF1-TRAF6). TRAF3, originally named CRAF1, interacts directly with the CD40 cytoplasmic tail through a region of similarity to the tumor necrosis factor-alpha (TNF-alpha) receptor-associated factors. TRAF3 binds only a single site, which contains the sequence PEQET, whereas TRAF1 and TRAF2 are capable of binding to either the PEQET site or an additional downstream domain.Specifications
| TRAF3 | |
| Polyclonal | |
| Unconjugated | |
| TRAF3 | |
| CAP 1, CAP1, CD40 binding protein, CD40bp, CRAF1, LAP1, LMP1 associated protein 1, TRAF3 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 7187 | |
| -20°C | |
| Liquid |
| Flow Cytometry, Immunoprecipitation, Western Blot | |
| 0.42 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q13114 | |
| TRAF3 | |
| TRAF3 Fusion Protein Ag12185 | |
| 20 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction