Learn More
Invitrogen™ TRAF2 Recombinant Superclonal™ Antibody (12HCLC)
Rabbit Recombinant Superclonal Antibody
Brand: Invitrogen™ 711288
Description
This antibody is predicted to react with Monkey, Horse, Pig and Dog Recombinant rabbit Superclonal™ antibodies are unique offerings from Thermo Fisher Scientific. They are comprised of a selection of multiple different recombinant monoclonal antibodies, providing the best of both worlds - the sensitivity of polyclonal antibodies with the specificity of monoclonal antibodies - all delivered with the consistency only found in a recombinant antibody. While functionally the same as a polyclonal antibody - recognizing multiple epitope sites on the target and producing higher detection sensitivity for low abundance targets - a recombinant rabbit Superclonal™ antibody has a known mixture of light and heavy chains. The exact population can be produced in every lot, circumventing the biological variability typically associated with polyclonal antibody production. Note: Formerly called Recombinant polyclonal antibody, this product is now rebranded as Recombinant Superclonal™ antibody. The physical product and the performance remain unchanged.
Tumor necrosis factor (TNF) receptor associated factors (TRAFs) were initially discovered as adaptor proteins that link the TNF receptor superfamily to signaling pathways and are thus important regulators of cell death and cellular response to stress. TRAF proteins share a homology region that allows them to bind to cell receptors and other TRAF proteins, causing the activation of different signal cascades depending on the TRAFs involved. For example, TRAF2 and TRAF3 directly bind to the CD40, a NF receptor superfamily member involved in inducing B cell immunity, and are critical for NF-kappa-B activation in mouse B lymphocytes. TRAF2 along with TRAF6 has also been shown to be required for CD40 signaling in nonhemopoietic cells. TRAF2 also interacts with the TRFR superfamily member lymphotoxin-beta receptor (LTbetaR) in association with TRAF3 and the apoptosis inhibitors cIAP1 and Smac.
Specifications
| TRAF2 | |
| Recombinant Superclonal | |
| 0.5 mg/mL | |
| PBS with 0.09% sodium azide; pH 7.2 | |
| P39429, Q12933 | |
| TRAF2 | |
| Peptides corresponding to Human TRAF2 (aa 483-501, 38-53). | |
| 100 μg | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
| Western Blot | |
| 12HCLC | |
| Unconjugated | |
| TRAF2 | |
| AI325259; E3 ubiquitin-protein ligase TRAF2; MGC:45012; OTTHUMP00000064745; RING-type E3 ubiquitin transferase TRAF2; TNF receptor associated factor 2; Tnf receptor-associated factor 2; TRAF2; TRAP; TRAP3; tumor necrosis factor type 2 receptor associated protein 3; tumor necrosis factor type 2 receptor-associated protein 3 | |
| Rabbit | |
| Protein A | |
| RUO | |
| 22030, 311786, 7186 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.