missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TRAF-5 Antibody (55A219), DyLight 680, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB600-1107FR
This item is not returnable.
View return policy
Description
TRAF-5 Monoclonal specifically detects TRAF-5 in Human samples. It is validated for Western Blot.
Specifications
| TRAF-5 | |
| Monoclonal | |
| DyLight 680 | |
| O00463 | |
| TRAF5 | |
| Anti-TRAF5 monoclonal antibody was raised against a fusion protein corresponding to amino acids 77 to 186 of human TRAF5. | |
| 0.1 mL | |
| Apoptosis, Signal Transduction, Zinc Finger | |
| 7188 | |
| Human | |
| Purified |
| Western Blot | |
| 55A219 | |
| Western Blot | |
| MGC:39780, RNF84RING finger protein 84, TNF receptor-associated factor 5 | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Primary | |
| Cross-reacts with Human. Not yet tested in other species. | |
| Store at 4C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction