missing translation for 'onlineSavingsMsg'
Learn More

TRAF-5 Antibody (55A219), DyLight 594, Novus Biologicals™

Product Code. 18408934 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18408934 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18408934 Supplier Novus Biologicals Supplier No. NB6001107DL594

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

TRAF-5 Monoclonal antibody specifically detects TRAF-5 in Human samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen TRAF-5
Applications Western Blot
Classification Monoclonal
Clone 55A219
Conjugate DyLight 594
Formulation 50mM Sodium Borate
Gene Alias MGC:39780, RNF84RING finger protein 84, TNF receptor-associated factor 5
Host Species Mouse
Immunogen Anti-TRAF5 monoclonal antibody was raised against a fusion protein corresponding to amino acids 77 to 186 of human TRAF5.
Purification Method Protein G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Apoptosis, Signal Transduction, Zinc Finger
Primary or Secondary Primary
Gene ID (Entrez) 7188
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.