missing translation for 'onlineSavingsMsg'
Learn More

TRAF-2 Antibody, Alexa Fluor™ 532, Novus Biologicals™

Product Code. 18403593 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18403593 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18403593 Supplier Novus Biologicals Supplier No. NBP177248AF532

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

TRAF-2 Polyclonal antibody specifically detects TRAF-2 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen TRAF-2
Applications Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Alexa Fluor 532
Formulation 50mM Sodium Borate
Gene Alias E3 ubiquitin-protein ligase TRAF2, EC 6.3.2.-, MGC:45012, TNF receptor-associated factor 2, TRAP3TRAP, tumor necrosis factor type 2 receptor associated protein 3, Tumor necrosis factor type 2 receptor-associated protein 3
Host Species Rabbit
Immunogen Antibody was raised against a 16 amino acid synthetic peptide from near the amino terminus of human TRAF2. The immunogen is located within amino acids 180 - 230 of TRAF2. Amino Acid Squence: CGKKKIPREKFQDHVK
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Apoptosis, Cancer, Cytokine Research, Zinc Finger
Primary or Secondary Primary
Gene ID (Entrez) 7186
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.