missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TRAF-2 Antibody (33A1293), DyLight 650, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB100-56715C
This item is not returnable.
View return policy
Description
TRAF-2 Monoclonal specifically detects TRAF-2 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| TRAF-2 | |
| Monoclonal | |
| DyLight 650 | |
| Q12933 | |
| TRAF2 | |
| Anti-TRAF2 monoclonal antibody was raised against a fusion protein corresponding to amino acids 205 to 222 of human TRAF2. | |
| 0.1 mL | |
| Apoptosis, Cancer, Cytokine Research, Zinc Finger | |
| 7186 | |
| Human | |
| Purified |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| 33A1293 | |
| Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin | |
| E3 ubiquitin-protein ligase TRAF2, EC 6.3.2.-, MGC:45012, TNF receptor-associated factor 2, TRAP3TRAP, tumor necrosis factor type 2 receptor associated protein 3, Tumor necrosis factor type 2 receptor-associated protein 3 | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Primary | |
| In HeLa, an approx. 57 kDa band is observed. | |
| Store at 4C in the dark. | |
| IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction