missing translation for 'onlineSavingsMsg'
Learn More

TRAF-2 Antibody (324522) [DyLight 680], Novus Biologicals™

Product Code. 30734963 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30734963 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30734963 Supplier Novus Biologicals Supplier No. FAB3277Y

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

TRAF-2 Monoclonal antibody specifically detects TRAF-2 in Human,Mouse,Rat samples. It is validated for Western Blot,Immunocytochemistry,Gene Knock-Out
TRUSTED_SUSTAINABILITY

Specifications

Antigen TRAF-2
Applications Western Blot, Immunocytochemistry, Gene Knock-Out
Classification Monoclonal
Clone 324522
Conjugate DyLight 680
Formulation 50mM Sodium Borate
Gene Alias E3 ubiquitin-protein ligase TRAF2, EC 6.3.2.-, MGC:45012, TNF receptor-associated factor 2, TRAP3TRAP, tumor necrosis factor type 2 receptor associated protein 3, Tumor necrosis factor type 2 receptor-associated protein 3
Host Species Mouse
Immunogen E. coli-derived recombinant human TRAF-2, Met1-Leu501, Accession # Q12933
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Apoptosis, Cancer, Cytokine Research, Zinc Finger
Primary or Secondary Primary
Gene ID (Entrez) 7186
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.