missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TRAF-1, Mouse anti-Human, Clone: TRAF1/3298, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-79791-20UG
This item is not returnable.
View return policy
Description
TRAF-1 Monoclonal specifically detects TRAF-1 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin, Protein Array.
Specifications
| TRAF-1 | |
| Monoclonal | |
| 0.2 mg/mL | |
| Immunohistochemistry 1-2 ug/ml, Immunohistochemistry-Paraffin 1-2 ug/ml, Protein Array | |
| Q13077, Q13077 | |
| TRAF1 | |
| Recombinant fragment of human TRAF-1 protein (around aa 73-219) (exact sequence is proprietary) | |
| 20 μg | |
| Apoptosis, Cancer, Cytokine Research | |
| 7185 | |
| Human | |
| Purified |
| Immunohistochemistry, Immunohistochemistry (Paraffin), Peptide Array | |
| TRAF1/3298 | |
| Unconjugated | |
| 10 mM PBS with 0.05% BSA | |
| EBI6MGC:10353, Epstein-Bar virus-induced protein 6, Epstein-Barr virus-induced protein 6, TNF receptor-associated factor 1 | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| This monoclonal antibody recognizes a protein of 52kDa, which is identified as TRAF-1 (TNFR-associated factor 1), which is highly useful for the differentiation of LyP and cALCL in patients with cutaneous CD30-positive lymphoproliferations. CD30-positive lymphoproliferations of the skin comprise 30% of all primary cutaneous T-cell lymphomas (CTCLs). Besides borderline cases this group includes lymphomatoid papulosis (LyP) and primary cutaneous anaplastic large T-cell lymphoma (cALCL). | |
| Store at 4°C. | |
| IgG2b κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction