missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ TRAF-1 Antibody (TRAF1/3299), Novus Biologicals™

Mouse Monoclonal Antibody

£178.00 - £351.00

Specifications

Antigen TRAF-1
Clone TRAF1/3299
Concentration 0.2 mg/mL
Applications ELISA
Classification Monoclonal
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
30043100
View Documents
Novus Biologicals™
NBP3-26740-100ug
100 μg
£351.00
100µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
30043966
View Documents
Novus Biologicals™
NBP3-26740-20ug
20 μg
£178.00
20µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

TRAF-1 Monoclonal antibody specifically detects TRAF-1 in Human samples. It is validated for ELISA
Specifications

Specifications

TRAF-1
0.2 mg/mL
Monoclonal
Purified
RUO
Human
EBI6MGC:10353, Epstein-Bar virus-induced protein 6, Epstein-Barr virus-induced protein 6, TNF receptor-associated factor 1
Recombinant fragment of human TRAF-1 protein (around aa 73-219) (exact sequence is proprietary)
Primary
Store at 4°C.
TRAF1/3299
ELISA
Unconjugated
Mouse
Apoptosis, Cancer, Cytokine Research
10 mM PBS with 0.05% BSA
7185
IgG1 κ
Protein A or G purified
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.