missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ TRAF-1 Antibody (TRAF1/3299) [DyLight 550], Novus Biologicals™

Product Code. 30065021 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30065021 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30065021 Supplier Novus Biologicals™ Supplier No. NBP327023R

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

TRAF-1 Monoclonal antibody specifically detects TRAF-1 in Human samples. It is validated for ELISA
TRUSTED_SUSTAINABILITY

Specifications

Antigen TRAF-1
Applications ELISA
Classification Monoclonal
Clone TRAF1/3299
Conjugate DyLight 550
Formulation 50 mM Sodium Borate
Gene Alias EBI6MGC:10353, Epstein-Bar virus-induced protein 6, Epstein-Barr virus-induced protein 6, TNF receptor-associated factor 1
Host Species Mouse
Immunogen Recombinant fragment of human TRAF-1 protein (around aa 73-219) (exact sequence is proprietary)
Purification Method Protein A or G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Apoptosis, Cancer, Cytokine Research
Primary or Secondary Primary
Gene ID (Entrez) 7185
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.