missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TRAF-1 Antibody (TRAF1/2770), Alexa Fluor™ 647, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-79924AF647
This item is not returnable.
View return policy
Description
TRAF-1 Monoclonal specifically detects TRAF-1 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| TRAF-1 | |
| Monoclonal | |
| Alexa Fluor 647 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| TRAF1 | |
| Recombinant fragment of human TRAF-1 protein (around aa 73-219) (exact sequence is proprietary) (Uniprot: Q13077) | |
| 0.1 mL | |
| Apoptosis, Cancer, Cytokine Research | |
| This monoclonal antibody recognizes a protein of 52kDa, which is identified as TNFR1 (TNFR-associated factor 1). CD30-positive lymphoproliferations of the skin comprise 30% of all primary cutaneous T-cell lymphomas (CTCLs). Besides borderline cases this group includes lymphomatoid papulosis (LyP) and primary cutaneous anaplastic large T-cell lymphoma (cALCL). Although the two entities overlap clinically, histopathologically, immunopathologically and genetically, they differ considerably in their prognosis. In particular, common feature of both cases is histologically the presence of atypical lymphoid CD30-positive T blasts and genetically a clonal T-cell-receptor rearrangement. However, both cases differ considerably in their clinical course: Lesions of LyP regress spontaneously, whereas those of cALCL persist and may progress and spread. Moreover, LyP patients do not benefit from an aggressive radio- and/or chemotherapeutic approach, in contrast to patients with cALCL. Besides, LyP and cALCL differ strongly in the expression of TRAF1 (tumor necrosis factor receptor (TNFR)-associated factor 1), a component of TNFR signaling: Whereas tumor cells of most LyP cases (ca. 84%) show a strong TRAF1 expression, tumor cells of cALCL reveal TRAF1 expression in only a few cases (ca. 7%). Antibody to TRAF1 is highly useful for the differentiation of LyP and cALCL in patients with cutaneous CD30-positive lymphoproliferations. | |
| Store at 4°C in the dark. |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| TRAF1/2770 | |
| Immunohistochemistry, Immunohistochemistry-Paraffin | |
| EBI6MGC:10353, Epstein-Bar virus-induced protein 6, Epstein-Barr virus-induced protein 6, TNF receptor-associated factor 1 | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction