missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human TRADD Antibody, R&D Systems™

Mouse Monoclonal Antibody
£167.00 - £371.00
Specifications
| Antigen | TRADD |
|---|---|
| Clone | 313203 |
| Concentration | LYOPH |
| Dilution | Intracellular Staining by Flow Cytometry 0.25 ug/10^6 cells, Immunocytochemistry 3-25 ug/mL, CyTOF-ready |
| Applications | Flow Cytometry, Immunocytochemistry, CyTOF |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18783368
![]() |
R&D Systems
MAB2658-SP |
25 μg |
£167.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18150091
|
R&D Systems
MAB2658 |
100 μg |
£371.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
TRADD Monoclonal specifically detects TRADD in Human samples. It is validated for Intracellular Staining by Flow Cytometry, Immunocytochemistry, CyTOF-ready.Specifications
| TRADD | |
| LYOPH | |
| Flow Cytometry, Immunocytochemistry, CyTOF | |
| Unconjugated | |
| Mouse | |
| Human | |
| Q15628 | |
| 8717 | |
| E. coli-derived recombinant human TRADD Met1-Ala312 Accession # Q15628 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Detects human TRADD in direct ELISAs and Western blots. In Western blots, no cross-reactivity with recombinant human (rh) BAFF R, rhTNF R1, rhHVEM, rhLT beta R, rhCD30, rhGITR, rh4-1BB, rhDR3, rhDR6, rhRANK, rhRELT, rhTAJ, rhEDAR, rhFas, recombinant mouse (rm) DC-TRAIL R1, rmDC-TRAIL R2, rhTRAIL R3, rhTRAIL R4, rhNGF R, rhCD27, rhTNF RII, rhTWEAK R, rhTRAIL R2, or rhCD40 is observed. |
| 313203 | |
| Intracellular Staining by Flow Cytometry 0.25 ug/10^6 cells, Immunocytochemistry 3-25 ug/mL, CyTOF-ready | |
| Monoclonal | |
| Purified | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| MGC11078, TNFRSF1A-associated via death domainTNFR1-associated DEATH domain protein, tumor necrosis factor receptor type 1 associated death domain protein, tumor necrosis factor receptor type 1-associated DEATH domain protein, tumor necrosis factor receptor-1-associated protein | |
| TRADD | |
| IgG2a | |
| Protein A or G purified from hybridoma culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title