Learn More
TRADD Mouse, anti-Human, Clone: 1C3H3, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 68573-1-IG-150UL
Description
The TRADD antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human TRADD. This antibody recognizes human, mouse, pig, rat antigen. The TRADD antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
| TRADD | |
| 1C3H3 | |
| Hs.89862, TRADD | |
| 150 μL | |
| 8717 | |
| Store at -20°C. Stable for one year after shipment. | |
| Liquid |
| Western Blot, ELISA | |
| 1000 μg/mL | |
| Mouse | |
| Primary | |
| Human, Mouse, Pig, Rat | |
| 150 kDa | |
| IgG1 |