missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TRACP/PAP/ACP5 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 1 publication
Brand: Novus Biologicals NBP2-41178
This item is not returnable.
View return policy
Description
TRACP/PAP/ACP5 Polyclonal specifically detects TRACP/PAP/ACP5 in Human, Mouse samples. It is validated for Western Blot, ELISA, Immunocytochemistry/Immunofluorescence.
Specifications
| TRACP/PAP/ACP5 | |
| Polyclonal | |
| Unconjugated | |
| PBS with 0.02% Sodium Azide | |
| ACP5 | |
| Antibody was raised against an 18 amino acid synthetic peptide near the carboxy terminus of human TRAP. The immunogen is located within the last 50 amino acids of TRAP. Amino Acid Squence: EASGKSLFKTRLPRRARP | |
| 0.1 mg | |
| Primary | |
| Human, Mouse | |
| IgG |
| Western Blot, ELISA, Immunocytochemistry, Immunofluorescence | |
| 1 mg/mL | |
| Western Blot 1 μg/mL, ELISA, Immunocytochemistry/Immunofluorescence 20 μg/mL | |
| HPAP, TRACP5a, TRACP5b, TRAP, TrATPase | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| 54 | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction