Description
- The PipettePilot™ app guides you through your microplate method to prevent errors
- Add traceability in experiments
- Easily capture notes, photos, and monitor your protocols
- Get immediate reproducibility
- Effortlessly create and share your protocols
- Compatibility with sciNote lets you structure your work into projects, experiments, and tasks, making it easy to visualize your work progress
- Access analytics and calibration history
- PipetteScope™ informs you exactly when your pipette needs to be calibrated
- Designed to communicate with PIPETMAN™ M Connected via Bluetooth™, the tablet interacts in real-time with the pipette and guides you through your protocol with PipettePilot™
- When complete, results can be saved, shared, and exported to sciNote, an open-source electronic lab notebook
Specifications
Specifications
| For Use With (Application) | PipettePilot™ application |
| For Use With (Equipment) | PIPETMAN™ M Connected Pipettes |
| Includes | SAMSUNG Galaxy Tab S3 with PipettePilot™ application, 5 V power adapter, tempered glass screen protector, tablet holder, microplate holder, microplate adapter, strip adapter, environment sensor, quick start guide |
| Warranty | 1 |