missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ TrackIt™ Ultra Low Range DNA Ladder
GREENER_CHOICE

Product Code. 15726548
Change view
Click to view available options
Quantity:
50 μg
Unit Size:
100 tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15726548 50 μg 100 tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15726548 Supplier Invitrogen™ Supplier No. 10488023
Limited-Time Offers on Selected Molecular Biology Essentials
Use promo code 27945
View all promotions
 Restricted Item
Estimated Shipment: 06-08-2026
Add to basket
Edge
Add to basket
This item is not returnable. View return policy

Includes

DNA Ladder and 1 vial 6X TrackIt™ Cyan/Yellow Loading Buffer

Ready-to-use DNA ladder for Low Range sample DNA analysis

Invitrogen TrackIt Ultra Low Range DNA Ladder is ready to use and designed for sizing and approximate quantification of double-stranded DNA in the range of 10 bp to 300 bp. TrackIt Ultra Low Range DNA Ladder consists of nine individual chromatography-purified DNA fragments and has a reference band at 100 bp for easy orientation.

TrackIt Ultra Low Range DNA Ladder is ideal for separation on 4–5% agarose gels.

Highlights of TrackIt Ultra Low Range DNA Ladder:

  • Sharp, clear bands—chromatography purified fragments for consistent and reliable results, stable at room temperature up to six months
  • Ready to use—premixed with 6X TrackIt Cyan/Yellow Loading Dye for tracking of DNA migration
  • Precise—an exact amount of DNA in each band

Product use

The double-stranded DNA ladder can be visualized on 4-5% agarose gels after ethidium bromide or SYBR Safe staining. The ladder is designed with a uniform intensity of DNA bands for a clear view of each band. An exact amount of DNA in each band allows approximate quantification of DNA samples.

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage • 500 μL TrackIt Ultra Low Range DNA Ladder
• 1 mL TrackIt Cyan/Yellow Loding Dye

Store at room temperature or at 4°C for up to 24 months. For longer periods, store at -20°C.
Concentration 0.1 μg/μL
Gel Type Agarose
Ready to Load Yes
Sample Loading Volume 1 mL
Size Range 10 to 300 bp
Volume (Metric) 500 μL
Gel Compatibility Agarose Gels
No. of Reactions 100 Applications
Quantity 50 μg
Shipping Condition Approved for shipment at Room Temperature or on Wet or Dry Ice
Technology Individual chromatography-purified DNA fragments
Product Line TrackIt
Product Type DNA Ladder
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.