missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
For the demanding requirements of high-temperature, simulated distillation analysis, choose the Thermo Scientific™ TRACE™ TR-SimDist Column. The column has a strongly cross-linked phase with an operating temperature limit of 400°C. Its stable low-bleed profile facilitates consistent background subtraction, which is common practice in these analyses.
Engineered for Superior Performance:
- Nonpolar phase, 100% dimethyl polysiloxane
- Optimized for simulated distillation analysis
- High temperature limits (400˚C for films <1.0μm; 370˚C for 2.65μm films)
- Strongly cross-linked
- High molecular weight hydrocarbons
- Simulated distillation
- ASTM D2887, D6532
- Simulated distillation
- Petroleum
- Xylenes
- DB-HT Sim Dis
- DB-2887
- BPX1
- Rtx™-2887
- HP-1
- Petrocol™ 2887
- Petrocol™ EX2887
Specifications
Specifications
| Max. Temperature | 370°C |
| Diameter (Metric) | 0.53 mm |
| Stationary Phase | Trace™ TR-SimDist |
| For Use With (Application) | GC/MS |
| Film Thickness | 0.10 μm |
| Length (Metric) | 10 m |
| Description | 10m x 0.53mm x 0.1um |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?