missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ TRACE™ TR-1 GC Columns

Use this non-polar GC column for method development and analysis of chlorinated and nitroaromatic compounds and environmental samples.

Brand:  Thermo Scientific™ 260A396P

Code : 16

Additional Details : Weight : 0.10000kg

 View more versions of this product

Product Code. 10592635

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.


Description

Description

Use the Thermo Scientific™ TRACE™ TR-1 GC column for method development and chlorinated and nitroaromatic compounds and environmental analyses. These non-polar phase, 100% dimethyl polysiloxane gas chromatography columns have a high operating temperature.

Specifications

Specifications

0.53 mm
TR-1
30 m
TRACE
3 μm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.