Learn More
Motic™ Trace Comparison Microscopy Software
Click to view available options
Type:
Trace Comparison Microscopy
Unit Size:
Each
Description
- Connect two or more Moticams or Motic™ Digital Microscopes to your computer and get immediate live comparison imaging.
- Cost effective.
- Overlay several live images, resize and independently rotate each image.
- Match two fingerprints with ease from live images or recall from a saved set.
- Check handwriting authenticity in a matter of seconds or determine whether two pieces of fabric belong to the same set.
Specifications
Specifications
| Type | Trace Comparison Microscopy |
| For Use With (Application) | Comparison microscopy, forensics |