Learn More
Novus Biologicals™ TRA2B Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand:  Novus Biologicals™ NBP3-30726
Product Code. 30048423
  • £432.00 / 100µg
Description
TRA2B Polyclonal antibody specifically detects TRA2B in Human,Mouse samples. It is validated for Immunoprecipitation,Western Blot
TRUSTED_SUSTAINABILITY
Specifications
Specifications
TRA2B
Polyclonal
Tris-citrate/phosphate pH 7 -8.
Rabbit
Affinity purified
RUO
6434
Store at 4°C. Do not freeze.
IgG
Immunoprecipitation, Western Blot
Unconjugated
DKFZp686F18120, hTRA2-beta, SFRS10, Splicing factor, arginine/serine-rich 10, splicing factor, arginine/serine-rich 10 (transformer 2 homolog, Drosophila), SRFS10, TRA-2 beta, TRA2-beta, TRAN2B, transformer 2 beta homolog (Drosophila), transformer 2 homolog, Transformer-2 protein homolog B, transformer-2 protein homolog beta, transformer-2-beta
Between 1 and 50
100 μg
Primary
Human, Mouse
Purified