missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TRA2B Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-49399
This item is not returnable.
View return policy
Description
TRA2B Polyclonal antibody specifically detects TRA2B in Human samples. It is validated for Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin)
Specifications
| TRA2B | |
| Polyclonal | |
| Immunohistochemistry 1:50 - 1:200, Immunocytochemistry/ Immunofluorescence 0.25-2 μg/mL, Immunohistochemistry-Paraffin 1:50 - 1:200 | |
| DKFZp686F18120, hTRA2-beta, SFRS10, Splicing factor, arginine/serine-rich 10, splicing factor, arginine/serine-rich 10 (transformer 2 homolog, Drosophila), SRFS10, TRA-2 beta, TRA2-beta, TRAN2B, transformer 2 beta homolog (Drosophila), transformer 2 homolog, Transformer-2 protein homolog B, transformer-2 protein homolog beta, transformer-2-beta | |
| This antibody was developed against a recombinant protein corresponding to amino acids: GYDRGYDDRDYYSRSYRGGGGGG | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
| Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS (pH 7.2), 40% Glycerol | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| 6434 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction